Fiber Optic Light Pipes
No support needed
  • Flexible arms offer positioning versatility
Self-supporting light pipes require no stand or additional support. They connect directly to the illuminator. Flexible gooseneck is constructed of glass fibers sheathed in a steel monocoil.
Illumination base (41720-40) sold separately.
64,500.00 - ₹80,300.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Light Pipe Length (in) Light Pipe Outside Dimension (OD) (in) Description
Availability Pricing
COMPARE
23 0.58 Self-Supporting Dual-Light Pipe, 23\" L Gooseneck
₹80,300.00INR / Each
COMPARE
23 0.46 Self-Supporting Light Pipe, Fiber Optic Dual
₹80,300.00INR / Each
COMPARE
20 0.58 Self-Supporting Single-Light Pipe, 20\" L Gooseneck
₹64,500.00INR / Each
COMPARE
20 0.56 Self-Supporting Light Pipe Fiber Optic Single
₹64,500.00INR / Each