Cole-Parmer® Extenders for 500- and 750-Watt Ultrasonic Processors
Use with tall or narrow vessels
  • Extenders are fabricated from titanium alloy TI-6AL-4V
  • Autoclavable
Extenders screw into threaded end probes of identical diameter in place of the replaceable tip. Recommended when working with tall narrow vessels such as Erlenmeyer flasks.
62,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Tip Diameter (cm) Tip Diameter (in) Description
Availability Pricing
COMPARE
1.27 0.5 Threaded Ultrasonic Probe Extender for 1/2\" Dia. Horn
₹62,000.00INR / Each
COMPARE
1.905 0.75 Threaded Ultrasonic Probe Extender for 3/4\" Dia. Horn
₹62,000.00INR / Each
COMPARE
2.54 1 Threaded Ultrasonic Probe Extender for 1\" Dia. Horn
₹62,000.00INR / Each