Extech Refrigerant Leakage Detector
See what you’re doing with handy LED light located right on probe tip
  • Color-coded lights visually indicate leak size
Detect refrigerant leaks in air conditioning and cooling systems with the Extech RD300 leak detector. A convenient LED work light at the tip of the probe illuminates dimly lit areas, making it easier to identify the source of a leak. Select from three sensitivity levels with multicolored light bars visually indicating the leak size. Mute button toggles the audible alarm on and off. Detector also features a flexible probe, low-battery indication, and auto-shutoff.
63,700.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Extech RD300 Refrigerant Leakage Detector WW-10112-25 Mfr # RD300
COMPARE
Refrigerant Leakage Detector
₹63,700.00INR / Each