This product is now discontinued, View Related Products
Extech Professional Digital Multimeters
Waterproof Rugged Design with CAT IV-600V safety rating for industrial applications
  • Double molded for waterproof (IP67) protection
  • 0.5% basic accuracy
  • Over-sized high contrast 4000 count LCD backlit display
  • 1000V input protection on all ranges
  • Dual sensitivity frequency functions
  • Relative Mode
  • Data Hold and Auto Power Off
  • Complete with double-molded test leads, and 9V battery.
23,100.00 - ₹27,800.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
AC Max (Volts) DC Max (Volts) True Rms
Availability Pricing
Extech EX503 Digital Multimeter 7.1, WW-26852-16 Mfr # EX503
COMPARE
1000 1000 Averaging
₹23,100.00INR / Each
Extech EX505 Digital Multimeter 7.1, WW-26852-18 Mfr # EX505
COMPARE
1000 1000 Yes
₹27,800.00INR / Each