Extech MO5X-Series Compact Moisture Meters
Take quick moisture level measurements with these pocket-sized meters
  • Convenient audible alert beeps faster as the moisture level increases
  • Feature built-in battery level check and measurement verification test
Quickly obtain moisture readings on wood, wall board, sheet rock, cardboard, plaster, concrete, and mortar—ideal for water damage restoration projects and testing walls, ceilings, tile, and under carpeting. Easy-to-read LCD shows both percent of moisture and low, medium, and high level icons. Meters also feature data hold, battery indicator, and auto power-off.
13,200.00 - ₹26,100.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Material Description
Availability Pricing
Extech MO50 Compact Pin Moisture Meter with Audible Alert WW-39756-02 Mfr # MO50
COMPARE
Wood moisture range: 5 to 50%; Other building materials: 1.5 to 33% Compact Pin Moisture Meter with Audible Alert
₹13,200.00INR / Each
Extech MO55 Compact, Combination Pin and Pinless Moisture Meter WW-59840-01 Mfr # MO55
COMPARE
Wood materials: 5 to 50% (pins), 0 to 99% (pinless); Other materials: 1.5 to 33% (pins), 0 to 99% (pinless) Compact, Combination Pin and Pinless Moisture Meter
₹26,100.00INR / Each