Extech IR260 Series Infrared Thermometers with Wide Temperature Range
No need to get dangerously close to high-temperature surfaces
  • Point-and-shoot noncontact design keeps you out of harm's way
  • Built-in laser improves aim ensuring measurement accuracy
  • Large backlit display is viewable in any lighting conditions
These handheld infrared (IR) thermometers are ideal for diagnostic and inspection applications where temperature is an indicator, especially for hard-to-reach areas and moving parts. Since IR temperature measurement does not require contact with surfaces, these thermometers reduce damage and contamination. These portable IR thermometers are easy to use—simply pull the trigger and the temperature will show on the display
17,700.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Temperature Input
Availability Pricing
Extech IR267 High Temperature Infrared Thermometer -58 to 1112F (-50 to 600C) with Type K Input (D:S 12:1) WW-90441-02 Mfr # IR267
COMPARE
1112 -58 Type K thermocouple
₹17,700.00INR / Each