Extech High Resolution Milliohm Meter
Provides resolution down to 0.01 mΩ
  • Built-in comparator offers Hi/Lo/Go resistance testing or selection
The benchtop meter provides high resolution down to 0.01 mΩ using 4-wire Kelvin clip test leads.It features seven ranges for wide 20mO to 20kO low resistance measurements, a large 0.8" digits 1999 count display, automatic zero, and 5V test voltage.
116,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) Resistance Max (MΩ) Resistance Min (KΩ)
Availability Pricing
Extech 380560 High Resolution Milliohm Meter; 110 VAC WW-20024-15 Mfr # 380560
COMPARE
110 20 20
₹116,000.00INR / Each