Extech Heavy-Duty Light Meters
Datalogging model allows the automatic or manual storing/recall of readings
These light meters offer wide measuring range up to 400,000 lux (40,000 fc), and provide cosine- and color-corrected capabilities. Meters feature a precision silicon photo diode and spectral response filter, and include relative mode indicating a change in light levels. Datalogging model (50512-32) automatically stores up to 16,000 readings or manually stores/recalls up to 99 readings.
85,100.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Data Logging
Availability Pricing
Extech HD450 Heavy-Duty Datalogging Light Meter, Model WW-50512-32 Mfr # HD450
COMPARE
16,000 continuous readings; 99 selected readings
₹85,100.00INR / Each