Extech EX65x Series True RMS 600A Clamp Meters
Get accurate readings when measuring distorted waveforms
  • Jaw size accommodates conductors up to 350 MCM
  • Data hold function freezes readings on display
  • LED worklight makes viewing in dimly lit areas easier
The 600A clamp meters feature True RMS for accurate readings when measuring distorted or noisy waveforms and a low impedance (LoZ) mode to prevent false readings caused by ghost voltages. The meters include a built-in noncontact voltage (NCV) detector for quick voltage detection, microampere function for HVAC flame rod current measurements, and a 6000-count backlit LCD.
31,400.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Extech EX655 True RMS 600A Clamp Meter with NCV; AC/DC WW-26848-03 Mfr # EX655
COMPARE
True RMS 600A Clamp Meter with NCV; AC/DC
₹31,400.00INR / Each