This product is now discontinued, View Related Products
Extech EasyView Thermohygrometers
Compact and rugged design featuring a large display
  • View results with unique bargraph
  • Available with datalogging and RS232 output
Measure temperature and humidity in laboratories, cleanrooms and storage areas with the remote probe. These thermohygrometers simultaneously displays any two parameters; temperature, humidity, dew point or wet bulb. View measurements in low light areas with backlit display. Meters feature switchable F/C units, MIN/MAX that will recall the highest and lowest readings, HOLD button, timer, elapsed time, programmable auto shutoff, low battery indicator and overrange indicators.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Battery Max RH Range (%) Max Temperature (° F)
Availability Pricing
Extech EA20 Easyview Thermohygrometer, 10 to 95% RH/, -4 to 140F WW-37803-03 Mfr # EA20
COMPARE
AAA Battery (6) 95 140
Call For Price
Extech EA25 Easyview Thermohygrometer with Datalogger WW-37803-04 Mfr # EA25
COMPARE
AAA Battery (6) 95 140