Extech Digital Strobotachometer
Analyze motion and speed – simply aim and synchronize flash rate with a rotating object
  • Handle provides convenient portability
This strobotachometer is ideal for measuring speeds of moving gears, fans, pumps, motors, and other equipment used in general maintenance, production, quality control, and laboratories. Exclusive microcomputer circuit and crystal control time-base produces high accuracy over a wide range.
59,900.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Flash Duration Max Flash Range (fpm) Power (VAC)
Availability Pricing
Extech 461830 Digital Strobotachometer, 115 VAC, 60 Hz WW-87700-09 Mfr # 461830
COMPARE
60 to 1000 µsec 10000
₹59,900.00INR / Each