This product is now discontinued, View Related Products
Eppendorf Multiporator Electroporation Systems
The Eppendorf® Multiporator® is optimally balanced for efficient and gentle electroporation of eukaryotic cells. The systems feature electronically regulated pulses in microsecond range to minimize damage to eukaryotic cells. The relevant parameters of voltage and pulse duration are directly set, and the electronic pulse discharge ensures that they will be maintained exactly — independent of the sample resistance — for reliable and reproducible results.
Applications include the transfection of plant cells.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
High Voltage Mode - Capacitance High Voltage Mode - Pulse Length High Voltage Mode - Voltage
Availability Pricing
Eppendorf 940000505 Eukaryotes (except yeast), 110-220 VAC, 60/50 Hz WW-36205-10 Mfr # 940000505
COMPARE
Call For Price
Eppendorf 940000602 Electroporation System, Bacteria Module; 110-220 VAC WW-36205-20 Mfr # 940000602