This product is now discontinued, View Related Products
Environmental Express StepSaver Extraction Stations
Individual glass stations for oil and grease analysis
  • Option of 47- or 90-mm glass funnels; or no funnel
The improved StepSaver™ is completely manufactured out of durable PVC and plastic materials. This compact, single-place station has a small footprint to save valuable benchtop space. These stations can be daisy-chained together to run multiple samples at once for a faster sample throughput.  The improved StepSaver still meets the EPA Method 1664 requirements for Oil and Grease extractions. 
132,000.00 - ₹204,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Material Description
Availability Pricing
COMPARE
Glass StepSaver Extraction Station, 47 mm, Glass Funnel; 1/Ea
₹157,000.00INR / Each
COMPARE
Glass StepSaver Extraction Station, 90 mm, Glass Funnel; 1/Ea
₹204,000.00INR / Each
COMPARE
Glass StepSaver Extraction Station, without Filter/Funnel; 1/Ea
₹132,000.00INR / Each