Environmental Express® Series 1500 Multiparameter Benchtop Meters
Optimized for laboratory measurements such as pH, ORP/mV, conductivity, resistivity, TDS, salinity, and temperature
  • Large monochrome LCD
  • Integrated electrode holder (up to 2 electrodes) that can be attached to either side of the meter
  • Simple user interface and single parameter display
  • 1000 Memory data points
  • Automatic Temperature Compensation (ATC) with temperature calibration
  • Adjustable auto shut-off time (1 to 30 minutes)
  • Auto stable, auto hold, and real time measurement modes with reading stability indicators
  • PC (standard USB) / printer (25 pin serial) connection via 2.5 mm diameter phono jack
Measurement parameters available vary by model. EC models measure conductivity, TDS, salinity, and resistivity. PC models measure pH, ORP/mV, conductivity, TDS, salinity, and resistivity. Users can choose the part number that best fits their needs. In addition to measurement parameters, each model can be bought as a kit or a meter only (meter only versions include the integrated electrode stand).
72,800.00 - ₹146,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
EC Benchtop Meter Kit
₹91,000.00INR / Each
COMPARE
EC Benchtop Meter with Electrode Stand
₹72,800.00INR / Each
COMPARE
PC Benchtop Meter Kit
₹146,000.00INR / Each
COMPARE
PC Benchtop Meter with Electrode Stand
₹102,000.00INR / Each