Environmental Express® Oakton® EC 100 Portable Conductivity Meter
Convenient, compact design with large display
  • Measured parameter and temperature shown on display at same time
  • Stability, calibration, and low-battery indicators
  • Waterproof and dust-resistant IP57 rating – great for use under harsh environments
The new addition to the Environmental Express Oakton product line features updates to measuring water quality easier and faster. The larger display is 2.75 in. by 2.0 in. for better viewing angle, and has indicators for battery life, stability, and calibration. It also shows both the parameter and temperature measurements at the same time. The EC 100 meter measures conductivity, TDS, salinity, and temperature.
The built-in microprocessor chip enables advanced functions, such as auto calibration, auto temperature compensation, parameter setting, self-diagnosis, auto power-off, and low-battery reminders. The meter uses advanced digital processing technology, intelligently improving the response time and accuracy of your measurements. Stable reading and auto lock display modes are available.
51,100.00 - ₹58,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Oakton EC 100 Portable Conductivity Meter with EC and ATC probes
₹51,100.00INR / Each
COMPARE
Oakton EC 100 Portable Conductivity Meter Kit with Case, EC and ATC probes, and Solutions
₹58,000.00INR / Each