Environmental Express® Method 5035 Field Sampling Kits
Extend sample hold time up to 14 days
  • All reagents and vials are certified and meet the requirements of EPA Method 5035
  • Keep vials upright and organized with the included plastic rack
  • Simply take your sample and place it in the vial—no field weighing required
These field sampling kits come with everything you need to obtain field samples for volatile organics in soil and waste testing. Convenient, economical, and easy-to-use—prelabeled vials, reagents, certifications, coring device, and instructions are included. All reagents and vials are certified with chromatograms available.
46,900.00 - ₹191,000.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Test Tests Per Kit Description
Availability Pricing
COMPARE
Volatile organics in soil and waste testing 1 SB Premium kit, 5mL MeOH in PTFE Tube
Call For Price
COMPARE
Volatile organics in soil and waste testing 1 SB Economy Kit, 5 mL MEOH in Vial
Call For Price
COMPARE
Volatile organics in soil and waste testing 1 SB Premium kit, 10 mL MeOH in PTFE Tube
Call For Price
COMPARE
Volatile organics in soil and waste testing 1 SB Economy Kit, 10 mL MEOH in Vial
Call For Price
COMPARE
Volatile organics in soil and waste testing 1 Organic-Free Premium kit, 5mL MEOH in PTFE Tube
Call For Price
COMPARE
Volatile organics in soil and waste testing 1 Organic-Free Economy kit, 5mL MEOH in Vial
Call For Price
COMPARE
Volatile organics in soil and waste testing 1 Organic-Free Premium kit, 10mL MEOH in PTFE Tube
Call For Price
COMPARE
Volatile organics in soil and waste testing 1 Organic-Free Economy kit, 10mL MEOH in Vial
Call For Price
COMPARE
Soil sampling 100 Soil Coring Device, 100 pk
₹46,900.00INR / Pkg of 1
COMPARE
Soil sampling 500 Soil Coring Device, 500 pk
₹191,000.00INR / Pkg of 1
COMPARE
20 Snip & Pour 10mL MeOH, 20 pk
Call For Price
Showing 1- 11 of 11