Environmental Express® Digestion Cups
Disposable digestion cups eliminate transfer steps
  • Cups that act as a digestion vessel or storage container and fit perfectly into the HotBlock and AutoBlock systems
  • Low metals background and EPA approved for digestions
  • Accessories include watch glasses, reflux caps, and transfer/storage racks
Approved for hot plate digestions and certified for low metals background.
10,400.00 - ₹35,800.00INR / Pkg of 500
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap Material Tube Material Volume (mL)
Availability Pricing
COMPARE
Polypropylene Polypropylene 15
₹35,800.00INR / Pkg of 1000
COMPARE
Polyethylene-foam lined polypropylene Polypropylene 100
₹10,400.00INR / Pkg of 225
Environmental Express SC545 Digestion Cup, PFA with PFA Cap, 50 mL; 1/Ea WW-53202-02 Mfr # SC545
COMPARE
PFA PFA 50
₹16,500.00INR / Each
Environmental Express SC543 Digestion Cup without Caps, PFA, 50 mL; 1/Ea WW-53202-03 Mfr # SC543
COMPARE
No cap included PFA 50
₹10,600.00INR / Each
COMPARE
No cap included Polypropylene 50
₹30,400.00INR / Pkg of 500
COMPARE
Polyethylene-foam lined polypropylene, green Polypropylene 50
₹30,400.00INR / Pkg of 500
COMPARE
Polyethylene-foam lined polypropylene, white Polypropylene 50
₹30,400.00INR / Pkg of 500
COMPARE
Unlined polypropylene Polypropylene 50
₹30,400.00INR / Pkg of 500
COMPARE
No cap included Polypropylene, rack lok 50
₹29,900.00INR / Pkg of 500
COMPARE
Polyethylene-foam lined polypropylene Polypropylene, rack lok 50
₹30,400.00INR / Pkg of 500
COMPARE
Unlined polypropylene Polypropylene, rack lok 50
₹30,400.00INR / Pkg of 500
COMPARE
Unlined polypropylene Polypropylene, clean 50
₹19,000.00INR / of 150
Showing 1- 12 of 12