This product is now discontinued, View Related Products
Environmental Express® Color-Coded Surrogates
Prevent costly mistakes by color coding your extraction fluids
  • Ideal for semi-volatile extractions like EPA Methods 3500C, 625, and 8270
  • Simply add 1 mL of the methanol based indicator to your sample
  • Store in your lab refrigerator
Color coded surrogates are methanol based and supplied in 25mL volumes in screw-top vials — add 1 mL to your sample and help avoid performing re-extractions. These surrogates act as an indicator to ensure the pH range of the sample is correct.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Volume (mL) Description
Availability Pricing
COMPARE
25 Color-Coded Surrogate, Acid (100 ug) for CLP; 25 mL
Call For Price
COMPARE
25 Color-Coded Surrogate, Acid (150 ug) for CLP; 25 mL
Call For Price
COMPARE
25 Color-Coded Surrogate, Base Neutral (100ug) for CLP; 25 mL