Environmental Express® BeO as Be Air Analysis Filters, Mixed Cellulose Ester (MCE), 37 mm
Ensure worker safety by monitoring workplace air quality
  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Traceable to NIST SRM 3105
  • Designed to meet QC requirements for NIOSH Method 7300 and US EPA Method SW-846
  • 37 mm diameter
  • 0.8 µm pore size
Beryllium Oxide is a toxic particulate material present in workplace samples that can be harmful to workers. Use these filters for method development or quality control checks to ensure worker safety. The source of the Beryllium Oxide applied to the filter is NIST SRM 1877 high-fired BeO powder. Blanks are supplied separately; contact us for information
17,900.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Concentration Element/Ion Description
Availability Pricing
COMPARE
25 µg/filter Beryllium Oxide as Beryllium MCE Air Analysis Filters, 25 µg/filter of BeO as Be
₹17,900.00INR / Each
COMPARE
10 µg/filter Beryllium Oxide as Beryllium MCE Air Analysis Filters, 10 µg/filter of BeO as Be
₹17,900.00INR / Each
COMPARE
5 µg/filter Beryllium Oxide as Beryllium MCE Air Analysis Filters, 5 µg/filter of BeO as Be
₹17,900.00INR / Each
COMPARE
2 µg/filter Beryllium Oxide as Beryllium MCE Air Analysis Filters, 2 µg/filter of BeO as Be
₹17,900.00INR / Each
COMPARE
1 µg/filter Beryllium Oxide as Beryllium MCE Air Analysis Filters, 1 µg/filter of BeO as Be
₹17,900.00INR / Each
COMPARE
0.5 µg/filter Beryllium Oxide as Beryllium MCE Air Analysis Filters, 0.5 µg/filter of BeO as Be
₹17,900.00INR / Each
COMPARE
0.2 µg/filter Beryllium Oxide as Beryllium MCE Air Analysis Filters, 0.2 µg/filter of BeO as Be
₹17,900.00INR / Each
COMPARE
0.1 µg/filter Beryllium Oxide as Beryllium MCE Air Analysis Filters, 0.1 µg/filter of BeO as Be
₹17,900.00INR / Each
Showing 1- 8 of 8