WW-59988-58 Mfr # 800 0161
COMPARE Manual or ultrasonic cleaning. Industrial use. 10 Ultrasonic Cleaner Detergent, Corrosion Remover; 10 L
5998858 WW-59988-59 Mfr # 800 0160
COMPARE Manual or ultrasonic cleaning. Industrial use. 25 Ultrasonic Cleaner Detergent, Corrosion Remover; 25 L
5998859 WW-59988-60 Mfr # 800 0101
COMPARE Manual or ultrasonic cleaning. Industrial use. 10 Ultrasonic Cleaner Detergent, Electronics; 10 L
5998860 WW-59988-61 Mfr # 800 0100
COMPARE Manual or ultrasonic cleaning. Industrial use. 25 Ultrasonic Cleaner Detergent, Electronics; 25 L
5998861 WW-59988-62 Mfr # 800 0131
COMPARE Manual or ultrasonic cleaning. Industrial use. 10 Ultrasonic Cleaner Detergent, Universal; 10 L
5998862 WW-59988-63 Mfr # 800 0130
COMPARE Manual or ultrasonic cleaning. Industrial use. 25 Ultrasonic Cleaner Detergent, Universal; 25 L
5998863 WW-59988-64 Mfr # 800 0073
COMPARE Manual or ultrasonic cleaning. Industrial or lab use. 10 Ultrasonic Cleaner Detergent, Gentle; 10 L
5998864 WW-59988-65 Mfr # 800 0029
COMPARE Manual or ultrasonic cleaning. Industrial or lab use. 25 Ultrasonic Cleaner Detergent, Gentle; 25 L
5998865 WW-59988-66 Mfr # 800 0055
COMPARE Manual or ultrasonic cleaning. Lab use. 20 Ultrasonic Cleaner Detergent, Surfactant-Free; 1 L, 20/Cs
5998866 WW-59988-67 Mfr # 800 0056
COMPARE Manual or ultrasonic cleaning. Lab use. 10 Ultrasonic Cleaner Detergent, Surfactant-Free; 2.5 L, 4/Cs
5998867 WW-59988-68 Mfr # 800 0057
COMPARE Manual or ultrasonic cleaning. Lab use. 10 Ultrasonic Cleaner Detergent, Surfactant-Free; 10 L
5998868 WW-59988-69 Mfr # 800 0058
COMPARE Manual or ultrasonic cleaning. Lab use. 25 Ultrasonic Cleaner Detergent, Surfactant-Free; 25 L
5998869 WW-59988-70 Mfr # 800 0075
COMPARE Manual or ultrasonic cleaning. Lab use. 20 Ultrasonic Cleaner Detergent, Foam-Inhibited; 1 L, 20/Cs
5998870 WW-59988-71 Mfr # 800 0076
COMPARE Manual or ultrasonic cleaning. Lab use. 10 Ultrasonic Cleaner Detergent, Foam-Inhibited; 2.5 L, 4/Cs
5998871 WW-59988-72 Mfr # 800 0077
COMPARE Manual or ultrasonic cleaning. Lab use. 10 Ultrasonic Cleaner Detergent, Foam-Inhibited; 10 L
5998872 WW-59988-73 Mfr # 800 0078
COMPARE Manual or ultrasonic cleaning. Lab use. 25 Ultrasonic Cleaner Detergent, Foam-Inhibited; 25 L
5998873 WW-59988-74 Mfr # 800 0095
COMPARE Manual or ultrasonic cleaning. Lab use. 20 Ultrasonic Cleaner Detergent, Mildly Acidic Foam-Inhibited; 1 L, 20/Cs
5998874 WW-59988-75 Mfr # 800 0096
COMPARE Manual or ultrasonic cleaning. Lab use. 10 Ultrasonic Cleaner Detergent, Mildly Acidic Foam-Inhibited; 2.5 L, 4/Cs
5998875 WW-59988-76 Mfr # 800 0097
COMPARE Manual or ultrasonic cleaning. Lab use. 10 Ultrasonic Cleaner Detergent, Mildly Acidic Foam-Inhibited; 10 L
5998876 WW-59988-77 Mfr # 800 0098
COMPARE Manual or ultrasonic cleaning. Lab use. 25 Ultrasonic Cleaner Detergent, Mildly Acidic Foam-Inhibited; 25 L
5998877