Electrothermal Wax Dispensers
Prevent wax blockage with heated tap
  • Accurate temperature control with digital display
  • Constructed of stainless steel with antimicrobial coating to inhibit bacteria growth
  • Enhanced safety with high temperature cut-off at 120°C
  • Prevent heat loss with fully insulated 7.5 L tank
This paraffin wax dispenser provides ultrafast heating for on-demand molten wax. Convenient “wax on tap” is available for an even flow of wax, ideal for your histology and pathology applications. The heated tap eliminates the possibility of wax blockage and the 0.5mm mesh filter screen prevents coarse particles from entering the tap. The nondrip lever has three settings — off, on-demand pull dispense, and continuous flow.
The set temperature can be controlled via the control panel, and allows for a temperature range ambient to 70°C. The digital display features a resolution of 1°C with a temperature accuracy of ±1°C. When set at 65°C, the unit will take approximately 90 minutes to fully melt 11 lbs (5 kg) of pelletized wax.
566,000.00 - ₹583,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (L) Power (VAC) Description
Availability Pricing
COMPARE
7.5 230 Electrothermal Wax Dispenser; 230V
₹566,000.00INR / Each
COMPARE
7.5 115 Electrothermal Wax Dispenser; 115V
₹583,000.00INR / Each