This product is now discontinued, View Related Products
Electrothermal Wax Dispenser
Eliminate wax solidifying at the tap
  • One-gallon capacity ensures ample supply of wax is on hand
  • Red neon light alerts you if an over-temperature condition occurs
  • Aluminum interior and exterior make cleaning easy
Wax dispenser easily heats wax to a liquid state for immediate ‘wax on tap’ for histology and pathology laboratories. Built-in thermostat regulates the wax temperature from ambient to 167°F (75°C). A special independent temperature control keeps dispenser tap heated to ensure wax always remains flowing when tap is open. Tap is a non-drip swivel lever with a replaceable filter. Four neon lights indicate power on, wax bath heater on, tap heater on, and wax bath overtemperature warning. Safety features include overtemperature cutout at 221°F (105°C) and nonresettable thermal fuse. Note: The dispenser is intended for paraffin wax use only with a melting range from 113 to 149°F (45 to 65°C) inclusive.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
COMPARE
230
Call For Price
COMPARE
115
Call For Price
COMPARE
230