Cole-Parmer® SW-200 Series Slide Warmer
Large capacity—holds up to fifty slides!
  • Temperature range from ambient to 100°C
  • Drying bars included for easier slide drying
  • Arrange slides in many different orientations
  • Dark surface makes it easy to see samples
Slide warmer facilitates tissue mounting and slide staining procedures, and is specifically designed for chemical, biological, and bacteriological laboratories. Dehydrate and set adhesives or perform non-adhesive section mountings on warming deck. Front panel has temperature adjustment dial and two lights — one for when power is on and one for when heater is on.
81,800.00 - ₹89,200.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Description
Availability Pricing
COMPARE
115 Microscope Slide Warmer, Holds 50 - 1\" x 3\" Slides; 115 VAC
₹89,200.00INR / Each
COMPARE
230 Microscope Slide Warmer, Holds 50 - 1\" x 3\" Slides; 230 VAC
₹81,800.00INR / Each