This product is now discontinued, View Related Products
Cole-Parmer® MP-250 Series Programmable Digital Melting Point Apparatus
Rotating viewing head ensures each user can analyze samples clearly
  • Set ramp rate between 0.2°C/min to 10.0°C in 0.1°C increment
  • RS-232 interface and USB ports allow for easy data transfer
  • Internal batch memory capacity of 1000 melts
  • View up to three samples simultaneously for greater efficiency
  • Height adjustable extension arm ensures proper viewing height for each user
  • Integral light and 8x magnifying lens enhance sample observation
  • Audible beeps indicate when temperature is stable and ready for sample
  • Extension arm folds away for easy storage
Use this programmable digital melting point apparatus to accurately measure, store, and transfer the melting points of your samples. Program the ramp rate from 0.2°C/minute to 10.0°C/minute in 0.1°C increments. Store up to four temperatures in temporary memory; batch memory permanently stores up to 1000 sets of results with date, time, and batch number. Output data to a computer or printer via RS-232 interface; a USB port allows data transfer to a flash drive.
Easily view three samples simultaneously through the 8x magnifying lens. An adjustable extension arm can be set to different viewing heights making it ideal for multiple users. A rotating viewing head and safety eye piece eliminate glare on the lens and protect you against hot zone. Conveniently located push button controls allow temperature recording without looking away from the sample. An ECO mode saves on energy use when keypad remains unused for a specified length of time.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Heat-Up Time Max Temperature (° C) Power (VAC)
Availability Pricing
COMPARE
30 minutes 400 120