This product is now discontinued, View Related Products
Electrothermal Omni OS6000 Series Reaction Stations
Simultaneously heat, stir, and reflux with one station—ideal for parallel synthesis
  • Control and maintain the same temperature and stirring rate at each position
  • Uniform heating across the heat block prevents “hot spots”
  • Compact footprint allows use under fume hood
  • Simple menus and touch pad interface make controlling quick and easy
Omni reaction stations are designed to provide easy and effective parallel synthesis and increase throughput in any chemistry lab. Heater cartridges heat to 428°F (220°C) with excellent stability of ±3°F (±1.5°C). Use with the included temperature probe to achieve temperature accuracy of ±0.9°F (±0.5°C). Unique airflow through the heater cartridge prevents outer casing from getting hot, reducing the risk of burns. Built-in magnetic stirrer provides stirring speed from 100 to 800 rpm.
Control each reaction vessel separately using a simple bidirectional open/close lever. Each vessel cap has a septum to allow sampling at each vessel position. Efficient condensing and refluxing is provided through the use of a liquid-cooled reflux head. Reflux/inerting head provides effective refluxing without the need for unwieldy condensing columns. A central gas port, combined with gas-tight PTFE caps, allows a vacuum to be pulled or an inert gas to be introduced. Connect a cooling plug and chiller to the cartridge to reach temperatures down to –22°F (–30°C) without the need for an ice bath.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Block - Tube / Vial Size Accepted (mm) Combination Block -Tube / Vial Size Accepted Power (VAC)
Availability Pricing
COMPARE
40.5 5 to 50 mL 230
Call For Price
COMPARE
40.5 5 to 50 mL 120
Call For Price
COMPARE
56.5 50 to 100 mL 230
Call For Price
COMPARE
56.5 50 to 100 mL 120
Call For Price
COMPARE
56.5 100 to 250 mL 230
Call For Price
COMPARE
56.5 100 to 250 mL 120