Cole-Parmer® CN-200 Series Analog Heating Controllers
Cost-effective control of resistive heating loads
  • Provide manual control of uncontrolled heating mantles, tapes, and cords
  • Compact—use on benchtop or mount to a frame
Use these external one-way controllers to manually regulate the power input to your laboratory heating equipment. Each heating controller features two indicators: "Power On" white neon light and "Mantle/Bunsen Heater On" amber neon light. As you turn the control dial clockwise to increase power, the amber indicator light begins to pulsate. Once maximum power is reached, the light will glow continually.
Heating controllers come with a short mains lead with an IEC socket for connecting to the resistive load. Easily place on benchtop or mount on a 1/2" (12.5 mm) diameter rod using the included support rod clamp.
44,600.00 - ₹79,800.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Control Output 1 Description
Availability Pricing
COMPARE
230 2300 W One-Way Analog Heating Controller, 2300 W; 230 VAC
₹71,900.00INR / Each
COMPARE
115 1100 W One-Way Analog Heating Controller, 1100 W; 115 VAC
₹79,800.00INR / Each
COMPARE
230 1800 W One-Way Analog Heating Controller, 1800 W; 230 VAC
₹45,300.00INR / Each
COMPARE
115 1150 W One-Way Analog Heating Controller, 1150 W; 115 VAC
₹44,600.00INR / Each
COMPARE
230 800 W One-Way Analog Heating Controller, 800 W; 230 VAC
₹50,700.00INR / Each
COMPARE
115 460 W One-Way Analog Heating Controller, 460 W; 115 VAC
₹50,800.00INR / Each