Cole-Parmer® HM-100 Series Uncontrolled Heating Mantles
Flexible heating element for efficient heat transfer and fast boiling times
  • Requires a power controller to operate
  • Exterior remains cool to the touch
  • Accepts round-bottom flasks
  • Maximum element temperature of 842°F (450°C)
These simple-to-use, low-cost heating mantles quickly heat up to 842°F (450°C). The replaceable coiled heating element suspended within a thermal insulating cartridge provides maximum heat transfer and support.
Powder-coated aluminum outer casing stays cool and is easy to clean. A stainless steel grounded earth screen and double fuses provided added safety. All heating mantles feature a support bracket for holding a support rod.
Note: A controller is required to operate the heating mantle—order separately.
63,600.00 - ₹84,300.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flask Size (mL) Power (VAC) Power (watts)
Availability Pricing
COMPARE
100 230 100
₹63,600.00INR / Each
COMPARE
500 230 280
₹83,800.00INR / Each
COMPARE
1000 230 380
₹84,300.00INR / Each
COMPARE
250 230 200
₹67,900.00INR / Each