Thermo Scientific® Electroporation Cuvettes, Sterile
High-quality disposable cuvettes are thoroughly cleansed to eliminate trace inhibitor materials
Each cuvette is sterile and individually packed with a presterilized transfer pipette. Color coded frosted caps enable trouble-free labeling and recognition. Sturdy polycarbonate construction withstands pulses of very high voltages or field strength and is autoclavable. Compatible with most electroporation systems.
60,900.00 - ₹61,300.00INR / Pkg of 50
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Description
Availability Pricing
Thermo Scientific 5510-11 Sterile Electroporation Cuvette, 1 mm gap width, white cap, 50/pack WW-25714-00 Mfr # 5510-11
COMPARE
Polycarbonate Sterile Electroporation Cuvette, 1 mm gap width, white cap, 50/pack
₹61,300.00INR / Pkg of 50
Thermo Scientific 5520 Sterile Electroporation Cuvette, 2 mm gap width, blue cap, 50/pack WW-25714-02 Mfr # 5520
COMPARE
Polycarbonate Sterile Electroporation Cuvette, 2 mm gap width, blue cap, 50/pack
₹61,300.00INR / Pkg of 50
Thermo Scientific 5540-11 Sterile Electroporation Cuvette, 4 mm gap width, green cap, 50/pack WW-25714-04 Mfr # 5540-11
COMPARE
Polycarbonate Sterile Electroporation Cuvette, 4 mm gap width, green cap, 50/pack
₹60,900.00INR / Pkg of 50