This product is now discontinued, View Related Products
Electroporation Buffers for Eukaryotic Cells
Provides quality transfection results
The best possible transfection results are obtained by using the original buffers from Eppendorf®, which have been tested for sterility as well as for the absence of mycoplasma, endotoxins, and pyrogens, and have undergone a thorough cytotoxicity test.
15,500.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Color Volume (mL) Description
Availability Pricing
Eppendorf 940002001 Electroporation Buffer for Eukaryotic Cells, Hypo-osmolar, 100 mL WW-36205-60 Mfr # 940002001
COMPARE
Clear 100 Electroporation Buffer for Eukaryotic Cells, Hypoosmolar, 100 mL
Call For Price
Eppendorf 940002109 Electroporation Buffer for Eukaryotic Cells, Iso-osmolar, 100 mL WW-36205-62 Mfr # 940002109
COMPARE
Clear 100 Electroporation Buffer for Eukaryotic Cells, Iso-osmolar, 100 mL
₹15,500.00INR / Each