This product is now discontinued, View Related Products
Electrix LED Linear Machine Light
LED bulb provides long life and low energy consumption
  • Rated IP67 for wet and dry locations
Direct mount LED linear machine light allows you to mount lighting wherever you need it. LED lighting means you won't have to change the bulbs often - bulb life is approximately 50,000 hours. Heavy duty aluminum housing is sealed for an IP67 rating. Can be mounted vertically or horizontally.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Length (")
Availability Pricing
Electrix LED Linear Machine Light, Direct Mount, 16" length WW-41310-37 Mfr # 8280.16
COMPARE
Call For Price
Electrix LED Linear Machine Light, Direct Mount, 24" length WW-41310-38 Mfr # 8280.24