Economical Ultraviolet Viewing Cabinets
Special contrast filter eliminates "blue haze" Models available for use in the lab or field
Use these economical viewing cabinets for gels and TLC plates in quality control, industrial, scientific, and viewing materials in lapidary applications. A special contrast control filter absorbs UV energy to protect your eyes and eliminate the "blue haze" often associated with UV viewing.
Removable bottom panel allows you to position cabinet over large or bulky samples or over a transilluminator. Cabinets include a lightweight curtain to block ambient light. Soft rubber viewport makes viewing comfortable. Cabinets measure 10-1/2"W x 12"H x 9"D. Shpg wt 4.0 lb (1.9 kg).
71,100.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (watts)
Availability Pricing
Analytik Jena 95007201 Chromato-Vue C-10 Mini UV Viewing Cabinet for Handheld 6W Lamps WW-97604-00 Mfr # 95007201
COMPARE
6
₹71,100.00INR / Each