Economical Dial Calipers
Wherever precision spatial measurements are needed, these easy-to-read dial calipers offer accurate measurements at an economical price. Made of fiberglass-reinforced plastic with beveled jaws.
  • Inside, outside, depth, and step measurements
  • Bar has inch and mm graduations on the front and a fractional conversion table on the back
  • No-glare lens for easy reading
  • Model 97100-00 is color-coded to highlight graduations of both 0.01" and 1/64"
  • Model 97100-10 has a metric readout that accurately reads to 0.001 mm
8,100.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max range (mm)
Availability Pricing
General Tools 144MM Analog/Dial Caliper, Plastic; 0 to 150 mm WW-97100-10 Mfr # 144MM
COMPARE
150
₹8,100.00INR / Each