This product is now discontinued, View Related Products
Ebro TFN Compact J/K/T/E Thermocouple Thermometers
Available in single- and dual- channel models
  • IP40-rated housing and keypad
These thermometers are constructed of ABS plastic and have an IP40 rating ideal for use in harsh environments. The use of an advanced internal microprocessor has selectable updating from 0.5 to 15 seconds produces an accuracy of ±0.5°F. The meters are multifunctional featuring max/min, Hold, °F/°C selectable units, adjustable sampling rate, large LCD, and auto power-off after inactivity to save on battery power. Available in single- and dual-channel, both capable of accepting any type of J, K, T, and E thermocouple probes with miniconnector. Dual-channel model also features T1, T2, and delta (T1–T2). Probes are sold separately.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Data Logging Number Of Probe Channels Types Of Probes Accepted
Availability Pricing
Ebro 1340-5522 TFN 520-Smp Compact E/J/K/T Thermocouple Thermometer with Single Input WW-91210-39 Mfr # 1340-5522
COMPARE
1 Type E, Type J, Type K, Type T
Call For Price
Ebro 1340-5532 TFN 530-Smp Compact E/J/K/T Thermocouple Thermometer with Dual Input WW-91210-40 Mfr # 1340-5532
COMPARE
2 Type E, Type J, Type K, Type T