This product is now discontinued, View Related Products
EBRO TFH Compact Digital Thermohygrometer
Ideal for use in harsh environments like labs, production and the food industry
  • IP40-rated humidity sensor
  • Compact size allows for convenient storage
These meters feature a robust impact-resistant ABS housing, degF/degC selection, user-defined measuring rate of 1 to 15 seconds, battery life indicator and an auto shutoff. A large LCD simultaneously displays RH and temperature readings.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Battery Max RH Range (%) Max Temperature (° F)
Availability Pricing
Ebro 1340-5610 (TFH 610) Tfh610 Compact Thermohygrometer (RH//Temperature) WW-37851-10 Mfr # 1340-5610 (TFH 610)
COMPARE
1.5V Lithium Battery (1) 100 122
Call For Price
Ebro 1340-5621(TFH620+TPH100) Compact Thermohygrometer with Remote Probe (RH//Temp/Dewith Wet) WW-37851-11 Mfr # 1340-5621(TFH620+TPH100)
COMPARE
1.5V Lithium Battery (1) 100 140