Eagle Overpack Salvage Plastic Drum Barrel
For secondary storage, containment, transport, or clean-up of 55-gallon drums or smaller containers
  • Passed a 3-PSI pressure test.
  • UN HM-181 approved
The salvage drums are constructed of yellow blow-molded high-density polyethylene (HDPE) with UV inhibitors, are durable and lightweight, as well as weather and chemical resistant. Ideal for safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration.
Drums are available with a metal band with bolt closure or screw top lids.
14,900.00 - ₹72,900.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Height (in) Description
Availability Pricing
Eagle 1695 Qverpack Salvage Plastic Drum Barrel, Metal Bolt Ring, 95 Gal WW-86110-02 Mfr # 1695
COMPARE
95 44 1/2 Overpack Salvage/Secondary Drum, 95 gallon
₹72,900.00INR / Each
Eagle 1650 Qverpack Salvage Plastic Drum Barrel, Screw-On Lid, 20 Gal WW-86110-04 Mfr # 1650
COMPARE
20 21 1/2 Overpack Salvage/Secondary Drum, 20 gallon
₹14,900.00INR / Each
Eagle 1661 Qverpack Salvage Plastic Drum Barrel, Screw-On Lid, 65 Gal WW-86110-06 Mfr # 1661
COMPARE
65 39 Overpack Salvage Drum, screw top, 65 gallon
₹37,800.00INR / Each