Eagle Modular Quick-Assembly Acid and Corrosive Safety Cabinets, HDPE
Assembles in minutes with no tools required
  • Stores corrosive liquids safety
  • Easy to store
Modular all-HDPE cabinets are designed to safely store corrosive or aggressive liquids in labs and industrial locations. Assembly is simple, and when not in use, cabinets can be broken down in minutes without tools and easily stored. The stand-alone cabinets can be assembled with the option of the lockable door opening to the right or left.
53,500.00 - ₹401,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Height (in) Number of Shelves
Availability Pricing
Eagle M04CRA Modular Quik-Assembly Acid and Corrosive Safety Cabinet, Benchtop, 2 Shelves, 4 Gal WW-81773-12 Mfr # M04CRA
COMPARE
4 22 2
₹102,000.00INR / Each
Eagle M04BEI Modular Quik-Assembly Safety Cabinet Top, 4 Gal., 1 Door, 2 Shelves - Beige WW-81773-18 Mfr # M04BEI
COMPARE
4 22 2
₹53,500.00INR / Each
Eagle M12CRA Modular Quik-Assembly Acid and Corrosive Safety Cabinet, 2 Shelves, 12 Gal WW-81773-20 Mfr # M12CRA
COMPARE
12 36 2
₹112,000.00INR / Each
Eagle M12Y Modular Quik-Assembly Safety Cabinet, 12 Gal., 1 Door, 2 Shelves - Yellow WW-81773-22 Mfr # M12Y
COMPARE
12 36 2
₹59,400.00INR / Each
Eagle M24CRA Modular Quik-Assembly Acid and Corrosive Safety Cabinet, 4 Shelves, 24 Gal WW-81773-28 Mfr # M24CRA
COMPARE
24 36 4
₹221,000.00INR / Each
Eagle M48CRA Modular Quik-Assembly Acid and Corrosive Safety Cabinet, 8 Shelves, 48 Gal WW-81773-30 Mfr # M48CRA
COMPARE
48 72 8
₹401,000.00INR / Each