Diversey™ Oxivir® Tb Disinfectant Cleaners & Wipes
Tough on pathogens, not on people
  • Disinfects surfaces in less than one minute
  • Ready-to-use one-step disinfectant saves you time
  • Non-irritating to skin and eyes
  • Gentle formula suitable for daily use
  • Penetrates through the toughest soils
  • Kills SARS-CoV-2 (COVID-19 virus)
  • Choose liquid solution or perforated wipes
Oxivir® Tb is a one-step disinfectant cleaner that is ready to use out of the bottle—no diluting needed. Simply wipe any hard, nonporous surface and allow the surface to remain wet for one minute. With no added phosphates, phenols, and fragrance, Oxivir Tb cleaner meets the lowest EPA toxicity category and is environmentally responsible – active ingredients break down into oxygen and water. Formulated with Accelerated Hydrogen Peroxide technology to deliver fast-acting, broad spectrum disinfection with enhanced cleaning power.
Disinfectant is effective against key pathogens such as TB, MSRA, Norovirus, and VRE. It kills HBV, HCV, and HIV in one minute; and is a potent tuberculocide and fungicide to effectively clean, disinfect, and deodorize.
Note: Food contact surfaces must be rinsed with potable water after using. Do not use on glassware, utensils, or dishes. If you are unsure of compatibility, it is recommended to test on a small, discreet surface.
2,400.00 - ₹523,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Applications Format Description
Availability Pricing
Diversey Oxivir® Tb Disinfectant Solution, 55 Gallon Drum WW-78900-86 Mfr # 101104053
COMPARE
General virucide, bactericide, tuberculocide, fungicide, sanitizer Clear, liquid Disinfectant Solution; 55 gallon drum
₹523,000.00INR / Each
Diversey Oxivir® Tb Disinfectant Solution, Four 1-Gallon Bottles WW-78900-87 Mfr # 100898636
COMPARE
General virucide, bactericide, tuberculocide, fungicide, sanitizer Clear, liquid Disinfectant Solution, four 1-gallon bottles
₹29,600.00INR / Case of 4
Diversey Oxivir® Tb Disinfectant Wipes, 11" x 12"; Case of 4 Canisters, 160 Wipes Ea WW-78901-02 Mfr # 5627427
COMPARE
General virucide, bactericide, tuberculocide, fungicide Wipes Disinfectant Wipes, 11\" x 12\"; Case of 4 Canisters, 160 Wipes Ea
₹51,100.00INR / Case of 4
Diversey Oxivir® Tb Disinfectant Solution; Case of 12 x 32 oz Bottles with Sprayers WW-78901-03 Mfr # 4277285
COMPARE
General virucide, bactericide, tuberculocide, fungicide, sanitizer Clear, liquid Disinfectant Solution; Case of 12 x 32oz Bottles with Sprayers
₹22,300.00INR / Case of 12
Diversey Oxivir® Tb Disinfectant Wipes, 6" x 7"; Case of 12 Canisters, 160 Wipes Ea WW-78901-04 Mfr # 4599516
COMPARE
General virucide, bactericide, tuberculocide, fungicide Wipes Disinfectant Wipes, 6\" x 7\"; Case of 12 Canisters, 160 Wipes Ea
₹50,200.00INR / Case of 12
Diversey Oxivir® Tb Disinfectant Solution; 1 Gallon Bottle WW-78901-05 Mfr # 100898636
COMPARE
General virucide, bactericide, tuberculocide, fungicide, sanitizer Clear, liquid Disinfectant Solution; one gallon bottle
₹7,400.00INR / Gallon of 1
Diversey Oxivir® Tb Disinfectant Wipes, 7" x 8"; Case of 12 Canisters, 60 Wipes Ea WW-78901-06 Mfr # 5388471
COMPARE
General virucide, bactericide, tuberculocide, fungicide, sanitizer Wipes Disinfectant Wipes, 7\" x 8\"; 12 Canisters, 60 Wipes Ea
₹28,200.00INR / Case of 12
Diversey Oxivir® Tb  Disinfectant Wipes, 6" x 7"; 160 Wipes Ea WW-78901-18 Mfr # 4599516
COMPARE
General virucide, bactericide, tuberculocide, fungicide Wipes Disinfectant Wipes, 6\" x 7\"; 160 Wipes Ea
₹4,200.00INR / Each
Diversey Oxivir® Tb Disinfectant Wipes, 7" x 8";  60 Wipes Ea WW-78901-19 Mfr # 5388471
COMPARE
General virucide, bactericide, tuberculocide, fungicide, sanitizer Wipes Disinfectant Wipes, 7\" x 8\"; 60 Wipes Ea
₹2,400.00INR / Each
Showing 1- 9 of 9