Diversey™ Avert® Sporicidal Disinfectant Wipes & Sprays
Cleans, disinfects, and deodorizes all in one step
  • Disinfects surfaces in one minute
  • Kills Clostridium difficile spores in four minutes
  • Ready-to-use one-step disinfectant saves you time
  • Effective against bacteria, antibiotic resistant bacteria, viruses, and fungi
Avert® is a ready to use, one-step, all-purpose disinfectant cleaner and deodorizer ideal for eliminating Clostridium difficile (C. diff) spores within four minutes on hard, nonporous surfaces such as glass, glazed porcelain, and stainless steel found in medical and healthcare facilities. Available in a spray solution or premoistened wipes, the bleach-based formula is also effective within a minute against various pathogens including bacteria, antibiotic resistant bacteria, viruses, and fungi.
Meets minimum active ingredient requirements for CDC, SHEA, APIC, and AORN.
58,800.00INR / Case of 12
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Applications Dilution Format
Availability Pricing
Diversey Avert® Sporicidal Disinfectant 6" x 7" Wipes; Case of 12 canisters, 160 wipes each WW-78901-09 Mfr # 100895790
COMPARE
Clostridium difficile spores, bacteria, antibiotic resistant bacteria, viruses, and fungi sanitizer Ready to use Wipes cannister
₹58,800.00INR / Case of 12