Disposable Hemacytometers
Precise fixed-depth counting chamber with superior accuracy and reproducibility
  • Single use
  • Light and unbreakable
  • No coverslips required
Our disposable hemocytometer is made of durable plastics used for manual cell counting. It consists of surface-patterned two enclosed 100µm height chambers with two ports for sample injection. It is a significant improvement of the conventional glass hemocytometer. Single usage reduces the risk of exposure to potential infectious material, increases productivity by eliminating cleaning and work interruptions.
33,400.00INR / Box of 1
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Disposable Hemacytometers 50/Box - Fuchs Rosenthal Grid WW-79003-02 Mfr # DHCF015
COMPARE
Disposable Hemacytometers 50/Box - Fuchs Rosenthal Grid
₹33,400.00INR / Box of 1