This product is now discontinued, View Related Products
Disposable Capsule Filters, Non-Sterile PTFE
These capsule filters are highly retentive and come in an all-polypropylene housing for excellent chemical compatibility and flow. Since no adhesives, binders, or surfactants are used in manufacturing, these capsules offer low filter extractables. The PTFE filter capsules are hydrophobic and ideal for use in critical applications or with aggressive chemicals such as acids, bases, and solvents.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Connections Effective Filtration Area (cm²) Pore Size (micron)
Availability Pricing
ZenPure Filter Capsule PTFE; 0.2um, 500cm2, 1/4"- 3/8" barb WW-49700-00 Mfr # MKPF020D2H2H
COMPARE
1/4\" to 3/8\" hose barb 500 0.2
Call For Price
ZenPure Filter Capsule PTFE; 1.0um, 500cm2, 1/4"- 3/8" barb WW-49700-02 Mfr # MKPF100D2H2H
COMPARE
1/4\" to 3/8\" hose barb 500 1
Call For Price
ZenPure Filter Capsule PTFE; 0.1um, 1250cm2, 1/4"- 3/8" barb WW-49700-10 Mfr # SKLF010S2H2H
COMPARE
1/4\" to 3/8\" hose barb 1250 0.1
Call For Price
ZenPure Filter Capsule PTFE; 0.2um, 1250cm2, 1/4"- 3/8" barb WW-49700-12 Mfr # SKLF020S2H2H
COMPARE
1/4\" to 3/8\" hose barb 1250 0.2
Call For Price
ZenPure Filter Capsule PTFE; 0.45um, 1250cm2, 1/4"- 3/8" barb WW-49700-14 Mfr # SKLF045S2H2H
COMPARE
1/4\" to 3/8\" hose barb 1250 0.45
Call For Price
ZenPure Filter Capsule PTFE; 1.0um, 1250cm2, 1/2" barb WW-49700-16 Mfr # SKLF100S4H4H
COMPARE
1/2\" hose barb 1250 1