Dilvac® Blue Enameled Steel Dewar Flasks with Handle and Lid
Safely transport liquid gases
  • Ideal for use with cold traps
  • Cushioned base prevents benchtop damage
  • Insulated lid and fold-down handle make it easy to transport
These economical Dewar flasks, featuring a blue enameled stainless steel outer container with a borosilicate glass inner flask, can safely be used with all liquid gases. Available in four capacity sizes, the Dewar flasks are specially designed for laboratory, hospital, and industrial use. The 1-, 2-, and 4.5-L models are also available with lid clamps.
52,400.00 - ₹224,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Blue Enameled Steel and Glass Dewar Flask with Handle and Lid, 1 L
₹52,400.00INR / Each
COMPARE
Blue Enameled Steel and Glass Dewar Flask with Handle and Lid, 2 L
₹61,700.00INR / Each
COMPARE
Blue Enameled Steel and Glass Dewar Flask with Handle and Lid, 4.5L
₹76,900.00INR / Each
COMPARE
Blue Enameled Steel and Glass Dewar Flask with Handle and Lid, 10 L
₹224,000.00INR / Each
COMPARE
Blue Enameled Steel and Glass Dewar Flask with Handle, Lid, and Lid Clamps, 1 L
₹57,400.00INR / Each
COMPARE
Blue Enameled Steel and Glass Dewar Flask with Handle, Lid, and Lid Clamps, 2 L
₹66,000.00INR / Each
COMPARE
Blue Enameled Steel and Glass Dewar Flask with Handle, Lid, and Lid Clamps, 4.5 L
₹83,500.00INR / Each
Showing 1- 7 of 7