This product is now discontinued, View Related Products
Traceable® Wireless Digital Thermometer Sets with Calibration
Read up to three separate modules up to 100 feet away without cables
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Transmitting modules have a remote probe to measure air, liquids, or solids
  • Monitors temperature continuously—data is transmitted every 30 seconds
Wireless signal from weatherproof remote sensor broadcasts temperature measurements up to 100 feet in an open area. Each set consists of a main unit/receiver and a remote module. A total of three modules may be used with the main receiver. The receiver displays data from all three transmitting modules along with temperature trends. Display MIN/MAX values at the touch of a button. Alarm sounds when temperature exceeds or falls below limits on any channel. Alarm sounds for one minute or until muted or disabled. °F or °C selectable. Supplied with hook-and-loop fasteners and bracket for wall mounting or has a flip-out stand for tabletop use.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (Outside) (° C) Max Temperature (Outside) (° F) Temperature Resolution (Outside) (° C)
Availability Pricing
COMPARE
70 158 0.1°C
Call For Price
COMPARE
60 140 0.1°C
Call For Price
COMPARE
60 140 0.1°C