This product is now discontinued, View Related Products
Traceable® Sentry Triple-Display Thermometers with Calibration
Simultaneously display high, low, and current temperature
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Monitors temperature continuously
Thermometers include a weatherproof remote sensor with a 10-ft. (3.05m) long 1/16"-diameter micro cable. The micro cable is ideal for applications that require a seal, such as a refrigerator, where a larger cable would interfere with the sealing of the door. Unplugging the external probe will activate an internal sensor for monitoring ambient temperature. Thermometer features a high-impact ABS plastic case, large LCD that updates every 10 seconds, and a simple one-button reset to clear the high/low memory. Supplied with hook-and-loop strip for wall mounting or has a flip-out stand for tabletop use.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Description
Availability Pricing
COMPARE
75 -51 Traceable® Sentry Triple-Display Thermometer with Calibration; -58 to 158F, Wire Probe
Call For Price
COMPARE
73 -50 Traceable® Sentry Triple-Display Thermometer with Calibration; -50 to 70C, Wire Probe
Call For Price
COMPARE
70 -50 Traceable® Sentry Triple-Display Thermometer with Calibration; -58-158F, Bottle Probe
Call For Price
COMPARE
74 -50 Traceable® Sentry Triple-Display Thermometer with Calibration; Celsius, Bottle Probe
Call For Price
COMPARE
71 -50 Traceable® Sentry Triple-Display Thermometer with Calibration; Fahrenheit, 5 mL Bottle
Call For Price
COMPARE
72 -50 Traceable® Sentry Triple-Display Thermometer with Calibration; Celsius, 5 mL Bottle