This product is now discontinued, View Related Products
Digi-Sense™ Traceable® Jumbo-Digit Atomic Wall Clocks with Calibration
Perfect size for benchtop use or wall mounting
  • Get accurate, reliable measurements with included NIST-traceable calibration
  • Large 3-3/4" (9.5 cm) digits are viewable from up to 100 feet (30.5 m) away
Compact case measures 9" wide by 5-3/4" high—perfect for benchtop use or wall mounting. Time signal which is regulated by an atomic clock is continuously transmitted from a corresponding location to automatically adjust clock to precise time and date. Calendar displays month/date and day of week. Clock is switchable for 12- or 24-hour time-of-day display. Clock can also be set to display ambient temperature in °F or °C.
Traceable® products are calibrated to NIST-traceable standards and ready to use as soon as you receive it. Each product goes through an intense vetting process before it can carry the Traceable name, and it includes NIST-traceable certification. This means that a 17025-accredited calibration test lab has confirmed that the product meets or exceeds all stated specifications, saving you both time and money by not having to calibrate separately.
NIST-traceable calibration also helps you meet quality standards and regulatory compliance for ISO, FDA, cGMP, VFC, CAP, CLIA and Joint Commission requirements.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Display Description
Availability Pricing
COMPARE
LCD, 3.75\" H digits Traceable® Jumbo-Digit Atomic Wall Clock with Calibration; US
Call For Price
COMPARE
LCD, 3.75\" H digits Traceable® Jumbo-Digit Atomic Wall Clock with Calibration; EU
Call For Price
COMPARE
LCD, 3.75\" H digits Traceable® Jumbo-Digit Atomic Wall Clock with Calibration; UK