Digi-Sense Thermowells - 304 Stainless Steel
Designed for easy replacement of your industrial probe without shutting the operation down.
To match your probe to a thermowell, first measure the distance from the probe’s tip to the top of the thread (typical thread length is 0.5" for 1/2" NPT fittings). Next, match this length to the probe depth of the thermowell (“A” dimension). Other thermowell sizes and materials are available—call our Application Specialists for more information.
Limited-Space Thermowell when you need a shorter insertion depth.
Standard Thermowell for general-purpose applications.
Heavy-Duty Thermowell for higher fluid velocity ratings.
3,200.00 - ₹28,800.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Description
Availability Pricing
COMPARE
1600 -32 Thermowell, 304 Stainless Steel, 2.5\" Length, 1/2\" Connection
₹3,200.00INR / Each
COMPARE
1600 -32 Thermowell, 304 Stainless Steel, 4\" Length, 1/2\" Connection
₹3,800.00INR / Each
COMPARE
1600 -32 Thermowell, 304 Stainless Steel, 6\" Length, 1/2\" Connection
₹5,000.00INR / Each
COMPARE
1600 -32 Thermowell, 304 Stainless Steel, 12\" Length, 1/2\" Connection
₹11,400.00INR / Each
COMPARE
1600 -32 Thermowell, 304 Stainless Steel, 4\" Length, 3/4\" Connection
₹4,200.00INR / Each
COMPARE
1600 -32 Thermowell, 304 Stainless Steel, 6\" Length, 3/4\" Connection
₹5,200.00INR / Each
COMPARE
1600 -32 Thermowell, 304 Stainless Steel, 9\" Length, 3/4\" Connection
₹8,700.00INR / Each
COMPARE
1600 -32 Thermowell, 304 Stainless Steel, 15\" Length, 3/4\" Connection
₹14,300.00INR / Each
COMPARE
1600 -32 Thermowell, 304 Stainless Steel, 18\" Length, 3/4\" Connection
₹17,700.00INR / Each
COMPARE
1600 -32 Thermowell, 304 Stainless Steel, 24\" Length, 3/4\" Connection
₹28,800.00INR / Each
Showing 1- 10 of 10