Digi-Sense Thermocouple Industrial Probes with Head and Thermowell
All-in-one, includes industrial probe head, element and thermowell
Thermocouple Probes Assemblies are ideal for use in industrial applications, chemical processes, production lines, ovens, furnaces, and vessels. The probes conductor is made of 20 gauge wire encased by ceramic insulators along with a standard size aluminum protection head. Head features a gasketed cap to seal out dust and moisture and houses a ceramic terminal block with screw terminals. Head has a 3/4" NPT (F) opening on the side for easy connection to electrical conduit. 316 stainless steel thermowells are included in assemble and are ideal for applications where removing of the probe without draining the vessel is necessary.
24,700.00 - ₹37,100.00INR / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Sheath material
Availability Pricing
COMPARE
1600
₹24,700.00INR / Each
COMPARE
1600
₹26,800.00INR / Each
COMPARE
1600
₹33,400.00INR / Each
COMPARE
1600
₹24,700.00INR / Each
COMPARE
1600
₹24,700.00INR / Each
COMPARE
1600
₹26,800.00INR / Each
COMPARE
1600
₹33,400.00INR / Each
COMPARE
1600
₹37,100.00INR / Each
COMPARE
1600
₹25,300.00INR / Each
COMPARE
1600
₹27,400.00INR / Each
COMPARE
1600
₹25,300.00INR / Each
COMPARE
1600
₹25,300.00INR / Each
COMPARE
1600
₹27,400.00INR / Each
Showing 1- 13 of 13