Digi-Sense Thermocouple Industrial Direct Insert Probe with Head
Available in spring loaded and non-spring loaded styles
Thermocouple Probes are ideal for use in industrial applications, chemical processes, production lines, ovens, furnaces, and vessels. The probes conductor is made of 18 gauge wire and is encased with Magnesium Oxide insulation, and has a 316 stainless steel sheath. Featuring a standard size aluminum protection head with a gasketed cap to seal out dust and moisture and houses a ceramic terminal block. Head has a 3/4" NPT(F) opening on the side for easy connection to electrical conduit.
Spring loaded probes: designed to bottom out when inserted in to a thermowell to produce an accurate reading. (Not designed for direct insertion in to medium)
Non-spring loaded probes: designed to be directly inserted into the medium being measured without the use of a thermowell.
13,500.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Sheath material
Availability Pricing
COMPARE
1330 316 Stainless Steel
₹13,500.00INR / Each
COMPARE
1330 316 Stainless Steel
₹13,500.00INR / Each
COMPARE
1330 316 Stainless Steel
₹13,500.00INR / Each
COMPARE
1650 316 Stainless Steel
₹13,500.00INR / Each
COMPARE
1650 316 Stainless Steel
₹13,500.00INR / Each
COMPARE
1650 316 Stainless Steel
₹13,500.00INR / Each
COMPARE
1650 316 Stainless Steel
₹13,500.00INR / Each
Showing 1- 7 of 7