Cole-Parmer® Thermocouple General-Purpose Surface Probes
Designed to measure the temperature at the surface without perpetrating the material
  • These probes are designed to measure surface temperature without penetrating the surface
  • Ideal for monitoring flat surfaces such as hot plates, furnaces, or molds
  • Response Time: 1.25 seconds or greater
  • Each probe comes with color coded ANSI coiled cable: type J-black, type K-yellow, type T-blue, and type E-purple.
  • Custom designs or changes to this product are available upon request.
Note the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
12,200.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type Description
Availability Pricing
COMPARE
500 T Type-T, Straight Surface Probe, 10\" L, Mini Conn, Exp, 5ft Coil Cord
₹12,200.00INR / Each
COMPARE
1200 K Type-K, Straight Surface Probe, 10\" L, Mini Conn, Exp, 5ft Coil Cord
₹12,200.00INR / Each
COMPARE
1200 J Type-J, Straight Surface Probe, 10\" L, Mini Conn, Exp, 5ft Coil Cord
₹12,200.00INR / Each