Cole-Parmer® Thermocouple General-Purpose Compact Probes
General-Purpose Probes Are Designed For Use with Liquids, Gases and Semisolids
  • Response Time: 2 seconds or greater.
  • Probe comes with PVC handle
  • Each probe comes with color coded ANSI miniconnectors: type J-black, type K-yellow, and type T-blue.
  • Custom designs or changes to this product are available upon request
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
6,400.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type Description
Availability Pricing
COMPARE
450 J Compact Thermocouple Probe, 4.5\" L, Grnd; Type J
₹6,400.00INR / Each
COMPARE
700 K Compact Thermocouple Probe, 4.5\" L, Grounded; Type K
₹6,400.00INR / Each
COMPARE
450 T Compact Thermocouple Probe, 4.5\" L, Grnd; Type T
₹6,400.00INR / Each