Digi-Sense Thermocouple Flexible Wire Probes, FEP-Insulated with Sealed Junction and Stripped Leads
FEP-insulated with sealed junction used for long-term immersion in semi-caustic environments
  • Each probe is hermetically sealed and tested to 25psi
  • Can be used for applications requiring an isolated junction
  • Response Time: 0.75 seconds or greater
  • All insulated-wire probes include straight insulated cable with no handle and stripped lead ends
  • Custom designs or changes to this product are available upon request
Do not subject type J exposed-junction probes to moisture as they will rust.
6,700.00 - ₹9,300.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type
Availability Pricing
COMPARE
400 K
₹6,700.00INR / Each
COMPARE
400 J
₹6,700.00INR / Each
COMPARE
400 T
₹6,700.00INR / Each
COMPARE
400 K
₹8,000.00INR / Each
COMPARE
400 J
₹8,000.00INR / Each
COMPARE
400 T
₹8,000.00INR / Each
COMPARE
400 K
₹9,300.00INR / Each
COMPARE
400 J
₹9,300.00INR / Each
COMPARE
400 T
₹9,300.00INR / Each
Showing 1- 9 of 9